Influenza A H3N2 (A/Brisbane/10/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Catalog # 103621-822
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H3N2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:ABW23353.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant hemagglutinin of Influenza A virus (A/Brisbane/10/2007 (H3N2)) comprises 526 amino acids with the predicted molecular mass of 59 kDa. As a result of glycosylation, it migrates as an approximately 90-100 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Ile531
- Shipping Temperature:Ambient
- Cat. No.:103621-822
Specifications
About this item
A DNA sequence encoding the extracellular domain of Influenza A virus (A/Brisbane/10/2007 (H3N2)) (ABW23353.1) hemagglutinin (Met 1-Ile 531) (Native, HA1+HA2, uncleaved) was expressed with a C-terminal polyhistidine.