Human Recombinant CCL26/Eotaxin-3 Protein (from Baculovirus Insect cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human CCL26 (NP_006063.1) (Met1-Leu94) was expressed.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_006063.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TSC-1|MIP-4alpha|MIP-4a|IMAC|SCYA26
- Protein/Peptide Name:CCL26
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human CCL26 consists of 71 amino acids and predicts a molecular mass of 8.4 kDa.
- Sequence:Met1-Leu94
- Shipping Temperature:Ambient