Cynomolgus/Rhesus Recombinant CD33/Siglec-3 Protein (His Tag) (from HEK293 Cells)
Catalog # 103585-616
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkeyRhesus monkey
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:EHH59833.1 / EHH30318.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:SIGLEC3|Siglec-3|p67
- Protein/Peptide Name:CD33
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant cynomolgus / rhesus CD33 comprises 244 amino acids and has a calculated molecular mass of 27.3 KDa. The apparent molecular mass of it is approximately 45.2 KDa in SDS-PAGE under reducing condition.
- Sequence:Met1-Gly248
- Shipping Temperature:Ambient
- Cat. No.:103585-616
Specifications
About this item
A DNA sequence encoding the cynomolgus / rhesus CD33 (EHH59833.1 / EHH30318.1) (Met1-Gly248) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD33 sequences are identical.