Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Recombinant ST6GAL1/CD75 Protein (His Tag) (from HEK293 Cells)
Rat Recombinant ST6GAL1/CD75 Protein (His Tag) (from HEK293 Cells)
Catalog # 103906-378
Supplier:  Sino Biological
CAS Number:  
Rat Recombinant ST6GAL1/CD75 Protein (His Tag) (from HEK293 Cells)
Catalog # 103906-378
Supplier:  Sino Biological
Supplier Number:  80866-R07H-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Gene ID:
    NP_001106815.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    St6gal|Siat1|St6galI|St6Gal-I|AW742324
  • Protein/Peptide Name:
    ST6GAL1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant rat ST6GAL1 consists of 396 amino acids and predicts a molecular mass of 46 kDa.
  • Sequence:
    Lys27-Cys403
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103906-378

Specifications

About this item

A DNA sequence encoding the rat ST6GAL1 (NP_001106815.1) (Lys27-Cys403) was expressed with a polyhistidine tag at the N-terminus.