Influenza A H5N1 (A/Egypt/1394-NAMRU3/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
103887-426
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H5N1
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant hemagglutinin of the Influenza A virus (A/Egypt/1394-NAMRU3/2007(H5N1)) consists of 521 amino acids and predicts a molecular mass of 59.7 kDa.
- Sequence:Met1-Ser525
- Shipping Temperature:Ambient
- Cat. No.:103887-426
A DNA sequence encoding the Influenza A virus (A/Egypt/1394-NAMRU3/2007(H5N1)) hemagglutinin (Met1-Ser525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.