Order Entry
Puerto Rico
ContactUsLinkComponent
Influenza A H5N1 (A/Cambodia/R0405050/2007) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect cells)
Influenza A H5N1 (A/Cambodia/R0405050/2007) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect cells)
Catalog # 103622-680
Supplier:  Sino Biological
CAS Number:  
undefined
Influenza A H5N1 (A/Cambodia/R0405050/2007) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect cells)
Catalog # 103622-680
Supplier:  Sino Biological
Supplier Number:  11710-V08B-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    H5N1
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method
  • Gene ID:
    ACI06178.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Hemagglutinin
  • Protein/Peptide Name:
    HA
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant hemagglutinin of Influenza A virus (A/Cambodia/R0405050/2007(H5N1)) consists 524 amino acids and predicts a molecular mass of 59.9 kDa.
  • Sequence:
    Met1-Gln529 (cleavage site mutations (GRRKKR to TETR))
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103622-680

Specifications

About this item

A DNA sequence encoding the Influenza A virus (A/Cambodia/R0405050/2007(H5N1)) hemagglutinin (ACI06178.1, with cleavage site mutations (GRRKKR to TETR)) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.