Influenza A H3N2 (A/Brisbane/10/2007) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Catalog # 103633-846
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H3N2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:ABW23353.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>97% as determined by SDS-PAGE
- Molecular Weight:The recombinant HA1 subunit of Influenza A virus H3N2 (A/Brisbane/10/2007 (H3N2)) hemagglutinin comprises 340 amino acids with the predicted molecular mass of 38 kDa. As a result of glycosylation, it migrates as an approximately 60-65 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Arg345
- Shipping Temperature:Ambient
- Cat. No.:103633-846
Specifications
About this item
A DNA sequence encoding the N-terminal segment of Influenza A virus H3N2 (A/Brisbane/10/2007 (H3N2)) hemagglutinin (ABW23353.1) (Met 1-Arg 345), termed as HA1, was fused with a polyhistidine tag at the C-terminus.