Human Recombinant Ephrin-A1/EFNA1 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103886-330
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:200 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Immobilized Ephrin-A1 Protein, Human, Recombinant (hFc Tag)(Cat:10882-H02H) at 2 μg/ml (100 μl/well) can bind EphA2 Protein, Mouse, Recombinant (His Tag)(Cat:50586-M08H), the EC50 is 13-64 ng/ml.
- Gene ID:NP_004419.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TNFAIP4|LERK-1|ECKLG|EFL1|B61|EPLG1|EFNA1|LERK1
- Protein/Peptide Name:EFNA1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human EFNA1 consists of 402 amino acids and predicts a molecular mass of 46.08 kDa.
- Sequence:Met1-Ser182
- Shipping Temperature:Ambient
- Cat. No.:103886-330
Specifications
About this item
A DNA sequence encoding the human EFNA1 (NP_004419.2) (Met1-Ser182) was expressed with the Fc region of human IgG1 at the C-terminus.