Cynomolgus Recombinant 4-1BBL/ CD137L Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103886-512
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:XP_015296398.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:AI848817|4-1BBL|4-1BB-L|Ly63l|Cd137l
- Protein/Peptide Name:TNFSF9
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant cynomolgus TNFSF9 consists of 445 amino acids and predicts a molecular mass of 47.8 kDa.
- Sequence:Arg68-Glu251
- Shipping Temperature:Ambient
- Cat. No.:103886-512
Specifications
About this item
A DNA sequence encoding the cynomolgus TNFSF9 (XP_015296398.1) (Arg68-Glu251) was expressed with the Fc region of human IgG1 at the N-terminus.