Order Entry
United States
ContactUsLinkComponent
 
Cynomolgus Recombinant Tie2/TEK Protein (Fc Tag) (from HEK293 Cells)
  103585-578
 :  
Cynomolgus Recombinant Tie2/TEK Protein (Fc Tag) (from HEK293 Cells)
  103585-578
 :  90292-C02H-100
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its binding ability in a functional ELISA.
    Immobilized mouse S1h-3C-mANGPT2 (Cat:50298-M07H) at 10 μg/ml (100 μl/well) can bind Cynomolgus TEK-Fc, The EC50 of Cynomolgus TEK-Fc is 0.7-1.64 μg/ml.
  • Gene ID:
    G7PS68
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    TIE-2|VMCM|TIE2|CD202B|VMCM1
  • Protein/Peptide Name:
    TEK
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant cynomolgus TEK is a disulfide-linked homodimer. The reduced monomer comprises 964 amino acids and has a calculated molecular mass of 107.6 KDa.
  • Sequence:
    Met1-Lys745
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103585-578

 

 

A DNA sequence encoding the cynomolgus TEK (G7PS68) (Met1-Lys745) was expressed with the Fc region of human IgG1 at the C-terminus.