Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Recombinant IFNGR/IFNGR1 Protein (Fc Tag) (from HEK293 Cells)
Rat Recombinant IFNGR/IFNGR1 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103666-418
Supplier:  Sino Biological
CAS Number:  
undefined
Rat Recombinant IFNGR/IFNGR1 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103666-418
Supplier:  Sino Biological
Supplier Number:  80182-R02H-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its binding ability in a functional ELISA. Immobilized Rat IFNGR1 hFc (Cat:80182-R02H) at 2 μg/ml (100 μl/well) can bind Rat Interferon Gamma hFc(Cat:80234-R02H), Biotinylated the EC50 of Rat Interferon Gamma hFc, Biotinylated is 60-300 ng/ml.
    2. Measured by its ability to inhibit rat IFNγ-mediated protection of L929 cells infected with vesicular stomatitisvirus (VSV). The ED50 for this effect is typically 0.5-4 ng/ml in the presence of 10 ng/ml of recombinant rat IFN-gamma.
  • Gene ID:
    Q6P6T3
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    IFNGR|IMD27A|IMD27B|CD119
  • Protein/Peptide Name:
    IFNGR1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant rat IFNGR/Fc is a disulfide-linked homodimer. The reduced monomer comprises 466 amino acids and has a predicted molecular mass of 52.4 kDa. The apparent molecular mass of the protein is approximately 117 and 65-80 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Ser241
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103666-418

Specifications

About this item

A DNA sequence encoding the rat IFNGR (Q6P6T3) (Met1-Ser241) was expressed with the Fc region of human IgG1 at the C-terminus.