Order Entry
Puerto Rico
ContactUsLinkComponent
Influenza A H1N1 (A/Hawaii/19/2007) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Influenza A H1N1 (A/Hawaii/19/2007) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Catalog # 103664-284
Supplier:  Sino Biological
CAS Number:  
Influenza A H1N1 (A/Hawaii/19/2007) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Catalog # 103664-284
Supplier:  Sino Biological
Supplier Number:  40508-V08H-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    H1N1
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Gene ID:
    ACA33723.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Hemagglutinin
  • Protein/Peptide Name:
    HA
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant HA1 subunit of the Influenza A virus (A/Hawaii/19/2007(H1N1)) hemagglutinin consists of 337 amino acids and predicts a molecular mass of 37.8 kDa.
  • Sequence:
    Met1-Arg343
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103664-284

Specifications

About this item

A DNA sequence encoding the Influenza A virus (A/Hawaii/19/2007(H1N1)) hemagglutinin (ACA33723.1) (Met1-Arg343), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.