Order Entry
Puerto Rico
ContactUsLinkComponent
Influenza A H5N1 (A/common magpie/Hong Kong/5052/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Influenza A H5N1 (A/common magpie/Hong Kong/5052/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Catalog # 103663-576
Supplier:  Sino Biological
CAS Number:  
undefined
Influenza A H5N1 (A/common magpie/Hong Kong/5052/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Catalog # 103663-576
Supplier:  Sino Biological
Supplier Number:  40044-V08H-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    H5N1
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Gene ID:
    ACJ26242.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Hemagglutinin
  • Protein/Peptide Name:
    HA
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    The secreted recombinant HA subunit of influenza A virus (A/common magpie/Hong Kong/5052/2007(H5N1)) comprises 525 amino acids and has a predicted molecular mass of 60 kDa.
  • Sequence:
    Met1-Gln530
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103663-576

Specifications

About this item

A DNA sequence encoding the influenza A virus (A/common magpie/Hong Kong/5052/2007(H5N1)) hemagglutinin (ACJ26242.1) (Met1-Gln530), termed as HA, was fused with a polyhistidine tag at the C-terminus.