Influenza A H5N1 (A/Egypt/2321-NAMRU3/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Catalog # 103622-314
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H5N1
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:ABP96850.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>97% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant influenza A H5N1 HA (A/Egypt/2321-NAMRU3/2007 (H5N1)) comprises 524 amino acids and has a predicted molecular mass of 60 kDa. As a result of glycosylation, it migrates as an approximately 65-80 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Gln531
- Shipping Temperature:Ambient
- Cat. No.:103622-314
Specifications
About this item
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza A H5N1 Hemagglutinin (A/Egypt/2321-NAMRU3/2007) (ABP96850.1) (HA1+HA2, uncleaved, with the cleavage site mutated (RRRKR-TETR) was expressed, fused with a C-terminal polyhistidine tag.