Influenza A H1N1 (A/Ohio/UR06-0091/2007) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Catalog # 103632-694
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H1N1
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:ABW40422.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>97% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant HA1 subunit of influenza A H1N1 HA (A/Ohio/UR06-0091/2007 (H1N1)) comprises 337 amino acids and has a predicted molecular mass of 37.8 kDa. As a result of glycosylation, the apparent molecular mass of the protein is approximately 55-70 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Arg343
- Shipping Temperature:Ambient
- Cat. No.:103632-694
Specifications
About this item
A DNA sequence encoding the N-terminal segment (Met 1-Arg 343) of the influenza hemagglutinin (A/Ohio/UR06-0091/2007 (H1N1)) (ABW40422.1), termed as HA1, was fused with a C-terminal polyhistidine tag.