ContactUsLinkComponent
TRIO.SETTLE Floor Stand Petri Plate Holder, Hardy Diagnostics
TRIO.SETTLE Floor Stand Petri Plate Holder, Hardy Diagnostics
Catalog # 76532-310
Supplier:  Hardy Diagnostics
undefined
TRIO.SETTLE Floor Stand Petri Plate Holder, Hardy Diagnostics
Catalog # 76532-310
Supplier:  Hardy Diagnostics
Supplier Number:  BAS368
Specifications
  • Description:
    Petri dish rack
  • Cat. No.:
    76532-310
Specifications
About this item
Floor-stand format culture media plate holder with an upper surface area to hold open media plate during 'passive' sampling, and a lower surface area to accommodate the lid of the media plate.
  • Extends to 1 meter
The system, fabricated in AISI 316 stainless steel, can be angled to direct the media plate’s exposure to the unidirectional airflow.