Cole-Parmer® Essentials Carboy with Spigot and Handle, Heavy Walled, Narrow-Mouth, HDPE, Antylia Scientific
Supplier: Antyila Scientific
Certificates
About this item
Heavy wall for added durability, allowing users to ship without worry.
- Can be used in sub-zero temperatures
- Effortless transport via heavy-duty swing handle
- Spigot quickly dispenses solutions
These narrow-mouth carboys are designed to provide a lifetime of service. Heavy-duty swing handles and thick walls offer safe and easy transport of solutions. The 3/4" BSP spigot pairs with a 12 mm deep thread in the carboy for leakproof performance. Approved for use with food products. Carboys are not autoclavable.