Order Entry
Puerto Rico
ContactUsLinkComponent
Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 594
Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 594
Catalog # 76485-360
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 594
Catalog # 76485-360
Supplier:  AAT BIOQUEST INC
Supplier Number:  25550
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Conjugation:
    iFluor® 594
  • Protein/Peptide Type:
    Native
  • Source:
    Eukaryotic cells
  • Species:
    Human
  • Size:
    1 mg
  • Protein Synonyms:
    WGA
  • Protein/Peptide Name:
    Wheat Germ Agglutinin
  • Cat. No.:
    76485-360

Specifications

About this item

Wheat germ agglutinin (WGA) is a lectin that binds to N-acetyl-D-glucosamine and sialic acid.

  • Glycoconjugate binding
  • Very bright signal
  • Multiple applications
Caution: For research use only.