Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
GTTR (Gentamicin Texas Red® labelled)

GTTR (Gentamicin Texas Red® labelled)

Supplier:   AAT Bioquest
GTTR (Gentamicin Texas Red® labelled)
Supplier:  AAT Bioquest
Certificates
GTTR  (Gentamicin Texas Red® labelled)
GTTR (Gentamicin Texas Red® labelled)

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Gentamicin is widely used for treating tuberculosis and Gram-negative infections and is particularly useful in neonatal intensive care units. Fluorophore-tagged drugs, such as gentamicin-Texas Red (GTTR) conjugates, have been used to identify drug trafficking routes across the BLB and into hair cells, and specifically in the cochlea - the sensory organ responsible for hearing. Identifying the mechanisms involved in the intracochlear trafficking of ototoxic drugs to hair cells is of fundamental as well as clinical importance. Texas Red-gentamicin conjugate (GTTR) provides researchers a valuable tool to study the cellular uptake, kinetics and distribution of gentamicin through fluorescence techniques, such as fluorescence microscopy, fluorescence correlation spectroscopy, single molecule spectroscopy. The TR fluorophore can be conveniently excited from 500-600 nm, with an emission maximum centered around 610 nm. Acute nephrotoxicity and permanent ototoxicity are serious side-effects that increase patient morbidity, or induce permanent auditory and vestibular deficits. Several approaches have been used to study the intracellular mechanisms induced by ototoxic drugs. However, ototoxic drugs must first cross the blood-labyrinth barrier (BLB) before entering sensory hair cells to exert their cytotoxic effect that leads to hearing loss and deafness. The BLB is similar to the blood-brain barrier (BBB), and is composed of tight junction-coupled endothelial and epithelial barrier layers that transport nutrients and ions from one side of the barrier to the other.

  • Well-cited conjugate
  • Useful for clinical studies
  • Easy-to-read signal
  • From Eukaryotic cells

Caution: For research use only.

Synonyms:  Gentamicin, Gentiomycin
  • CAS Number:1403-66-3
  • Formula:C₂₁H₄₃N₅O₇
  • Molecular Weight:477.603 g/mol
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Vial
    100 µg
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $237.68
    $237.68
    Each (100µG)
       $237.68

    Supplier Number: 24300
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $1,142.32
    $1,142.32
    Each (1mg)
       $1,142.32

    Supplier Number: 24301
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.