Order Entry
Puerto Rico
ContactUsLinkComponent
Native Amanita phalloides Phalloidin (from Eukaryotic cells), iFluor® 514
Native Amanita phalloides Phalloidin (from Eukaryotic cells), iFluor® 514
Catalog # 76483-874
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Native Amanita phalloides Phalloidin (from Eukaryotic cells), iFluor® 514
Catalog # 76483-874
Supplier:  AAT BIOQUEST INC
Supplier Number:  23116
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Conjugation:
    iFluor® 514
  • Protein/Peptide Type:
    Native
  • Source:
    Eukaryotic cells
  • Species:
    Amanita phalloides
  • Size:
    300 tests
  • Protein Synonyms:
    Amanita phalloides
  • Protein/Peptide Name:
    Phalloidin
  • Cat. No.:
    76483-874

Specifications

About this item

This yellow fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 514-labeled phalloidin) selectively binds to F-actins.

  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Caution: For research use only.