Order Entry
Puerto Rico
ContactUsLinkComponent
 
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 405
  76483-870
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 405
  76483-870
 :  23111
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pack type:
    Vial
  • Conjugation:
    Alexa Fluor® 405
  • Protein/Peptide Type:
    Native
  • Source:
    Eukaryotic cells
  • Species:
    Amanita phalloides
  • Size:
    300 tests
  • Protein Synonyms:
    Amanita phalloides
  • Protein/Peptide Name:
    Phalloidin
  • Cat. No.:
    76483-870

 

 

This blue fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 405-labeled phalloidin) selectively binds to F-actins.

  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
 : For research use only.