VWR® Single Channel Switchable Electronic Timer
Product Details & Documents
This single-channel timer is switchable between 19 hours, 59 minutes and 99 minutes, 59 seconds. It features a 3-key operation for ease of use.
- Count direction: up/down
- Last time set memory
- Audible alarm; displays elapsed time after countdown
Mounting options: lay flat on bench, flip-open stand for vertical view, and a magnetic back for metal surface attachment.
Certifications: Certificate of calibration and individual serial number, these electronic timers are tested and calibrated in an exclusive triple-accredited/-registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: Supplied with fold-away stand, pocket clip, magnetic back, one AAA battery, and certificate of calibration. Includes a statement of traceability indicating accuracy to NIST.
Specifications
- Description:Digital timer, single channel
- W×H×D:64×15.2×64 mm
- Accuracy:±5 s/day
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



