To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Clone:IgG
- Conjugation:trFluor™ Eu
- Host:Donkey
- Isotype:IgG (H, L)
- Reactivity:Goat
- Size:50 µg
- Environmentally Preferable:
- Cross Adsorption:Min × Human
- Cat. No.:76484-762
- Packaging:Vial
Specifications
About this item
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- High stability and emission yield
- Low background signal
- Large stokes shift
- Long emission half-life
- Multiple applications