Human Recombinant IL18BP Protein (Fc Tag) (from HEK293 Cells)
103905-956
:
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Measured by its ability to inhibit the IL-18-induced response of KG‑1 human acute myelogenous leukemia cells. The ED50 for this effect is 0.1‑0.6 µg/ml in the presence of 50 ng/ml of recombinant human IL-18.
- Gene ID:AAD17187.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Igifbp|IL-18BP|MC54L
- Protein/Peptide Name:IL18BP
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human IL18BP consists of 402 amino acids and predicts a molecular mass of 44.4 kDa.
- Sequence:Met1-Gly192
- Shipping Temperature:Ambient
- Cat. No.:103905-956
A DNA sequence encoding the human IL18BP (AAD17187.1) (Met1-Gly192) was expressed with the Fc region of human IgG1 at the C-terminus.