Order Entry
Puerto Rico
ContactUsLinkComponent
Native Biotin (from Eukaryotic cells), Cal-520®
Native Biotin (from Eukaryotic cells), Cal-520®
Catalog # 76483-096
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Native Biotin (from Eukaryotic cells), Cal-520®
Catalog # 76483-096
Supplier:  AAT BIOQUEST INC
Supplier Number:  20605
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Cal-520®
  • Protein/Peptide Type:
    Native
  • Source:
    Eukaryotic cells
  • Size:
    5×50 µg
  • Protein Synonyms:
    Vitamin H, B7
  • Protein/Peptide Name:
    Biotin
  • Cat. No.:
    76483-096

Specifications

About this item

Biotin and biocytin bind with high affinity to both avidins and streptavidins.

  • High specificity
  • Sensitive detection
  • Multiple applications
Caution: For research use only.