Order Entry
Puerto Rico
ContactUsLinkComponent
CosMCPrep Plasmid Purification Kit, Beckman Coulter
CosMCPrep Plasmid Purification Kit, Beckman Coulter
Catalog # 76177-115
Supplier:  Beckman Coulter
CAS Number:  
undefined
CosMCPrep Plasmid Purification Kit, Beckman Coulter
Catalog # 76177-115
Supplier:  Beckman Coulter
Supplier Number:  A29174
CAS Number:  

Specifications

  • Sample type:
    Cultured cells
  • Description:
    CosMCPrep plasmid purification kit
  • Size:
    4000 preparations
  • Cat. No.:
    76177-115
  • Format:
    Liquid
  • Application:
    Plasmid purification

Specifications

About this item

A SPRI paramagnetic bead-based plasmid purification system that uses a single protocol to purify a variety of high and low copy number template types. This flexible system is easily automated without sacrificing product quality.

  • Automated available

The single protocol, ease of automation and high quality results make CosMCPrep an ideal solution for all your plasmid purification needs. Kit components: L2 buffer, N3 buffer, pur4 buffer and RE1 buffer for 4000 preparations.