USP Standards, 100 µg/ml, Spex CertiPrep®
Supplier: SPEX CERTIPREP LLC
Certificates
About this item
Tests and specifications for the safety of botanical products.
- Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
- Certified to DQS to ISO 9001:2015
The United States Pharmacopeia (USP) creates standards for the monitoring of pharmaceutical and botanical products. General Chapter <561> Articles of Botanical Origin, outlines tests and specifications for the safety of botanical products. The chapter outlines sampling procedures and tests for general characteristics, such as for moisture, ash, starch and fiber. The chapter also describes methods for extractables and volatile compounds. In the interests of safety, USP <561> has procedures for the testing of mycotoxins and designates targeted pesticides. SPEX CertiPrep provides standards for testing of mycotoxins and pesticides under the General Chapter <561>.
Certifications: Supplier Certificate of Analysis (CoA) is included with product shipment.