Mycotoxins Standard Mix, 10 µg/ml, Spex CertiPrep®
Catalog # SXMYCO-1
Supplier: SPEX CERTIPREP LLC
CAS Number:
Specifications
- Concentration:10 µg/ml
- Matrix:Acetonitrile
- Description:Mycotoxin Standard Mix
- Size:1 ml
- Storage Conditions:Freezer
- Cat. No.:SXMYCO-1
- Contents:Aflatoxin B1, Aflatoxin B2, Aflatoxin G1, Aflatoxin G2, Ochratoxin
Specifications
About this item
Test for mycotoxins to help prevent foodborne illnesses and significant crop losses.
- Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
- Certified to DQS to ISO 9001:2015
- Mix includes 5 of the most commonly tested mycotoxins
- 10 ppm mycotoxin mix in acetonitrile matrix
- 1 ml ampule
Mold and fungi are all over the world and are of major concern in agricultural products. Fungi and their spores can spread throughout crops and food stores tainting crops. Fungi can proliferate into colonies with the right environmental conditions which destroy crops by consuming them or poison them with toxic secondary metabolites called mycotoxins.
Certifications: Mycotoxins are ISO 17034 accredited. Supplier Certificate of Analysis (CoA) is included with product shipment.
Caution: Flammable, Toxic, Irritant, Health Hazard