Oxivir® Tb Ready To Use Disinfectant Cleaner, Diversey
Supplier: DIVERSEY INC.
Certificates
About this item
Delivers fast, effective cleaning performance.
- Virucide, bactericide, tuberculocide, fungicide and sanitizer
- Effective against key pathogens
- Kills HBV, HCV and HIV in 1 minute
- Tuberculocidal in 5 minutes
- Fungicidal in 3 minutes
- No VOCs, no added fragrance, no NPEs
- Environmentally responsible, active ingredient breaks down to oxygen and water
Ready-to-use disinfectant cleaner based on proprietary hydrogen peroxide (AHP®) technology to deliver fast, effective cleaning performance. Disinfects in 60 seconds. Virucide, bactericide, tuberculocide, fungicide and non-food contact sanitizer. Kills MRSA and Norovirus. Meets bloodborne pathogen standards for decontaminating blood and body fluids. Colorless with a characteristic scent.