Covidyte™ ED450 is a peptide substrate containing 12 amino acid sequence (VNSTLQSGLRKM) that can be cleaved by coronavirus proteases. The dark-FRET peptide contains Dabcyl (quencher) and Edans (donor) on the C-and N-terminals respectively where the fluorescence of Edans is effectively quenched by Dabcyl when the peptide is intact. When the peptide is hydrolyzed by coronavirus proteases, the Edans fragment generates significantly enhanced fluorescence since its fluorescence is no longer quenched by Dabcyl. The activity of coronavirus proteases can be effectively monitored by the fluorescence intensity of Edans. Covidyte™ ED450 is a convenient tool for screening inhibitors of coronavirus proteases.
- MW 2034.36
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Source:
- Species:
- Environmentally Preferable: