Cell Meter™ Intracellular Fluorimetric Hydrogen Peroxide Assay Kit *Blue Fluorescence Optimized for Flow Cytometry*, AAT Bioquest
76481-934
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Description:Cell Meter™ intracellular fluorimetric hydrogen peroxide assay kit *Blue Fluorescence Optimized for Flow Cytometry*
- Cat. No.:76481-934
- No. of tests:100
Hydrogen peroxide is a reactive oxygen metabolic by-product that serves as a key regulator for a number of oxidative stress-related states.
- Robust protocol
- High sensitivity
- Quantifies h2o2 in live cells
- Optimized for flow cytometry
: For research use only.