MegaWox™ polyHRP-Protein A Conjugate, AAT Bioquest
Catalog # 76485-250
Supplier: AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:MegaWox™ polyHRP-Protein A Conjugate
- Size:1 mg
- Cat. No.:76485-250
Specifications
About this item
Protein A is a 42-kDa protein found in the cell wall of Staphylococcus aureus.
- No unconjugated hrp
- Highly purified
- Compatible with all major substrates
- Multiple applications
- High sensitivity
Caution: For research use only.