ContactUsLinkComponent
Signal Guard™ HRP Conjugate Stabilizer, AAT Bioquest
Signal Guard™ HRP Conjugate Stabilizer, AAT Bioquest
Catalog # 76481-882
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Signal Guard™ HRP Conjugate Stabilizer, AAT Bioquest
Catalog # 76481-882
Supplier:  AAT Bioquest
Supplier Number:  11010
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Signal Guard™ HRP conjugate stabilizer
  • Size:
    50 ml
  • Cat. No.:
    76481-882
Specifications
About this item
The major advantage of a liquid HRP-conjugate is that it eliminates the chance for human error when dissolving or diluting the lyophilized or concentrated conjugate.
  • Permits liquid-hrp conjugates to be stored at 4°c for 24 months or RT for 6 months
  • Can create stock solutions or ready-to-use reagents
Caution: For research use only.