ContactUsLinkComponent
ReadiUse™ TCEP Removal Solution, AAT Bioquest
ReadiUse™ TCEP Removal Solution, AAT Bioquest
Catalog # 76484-212
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ReadiUse™ TCEP Removal Solution, AAT Bioquest
Catalog # 76484-212
Supplier:  AAT Bioquest
Supplier Number:  5540
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    ReadiUse™ TCEP removal solution
  • Size:
    1 ml
  • Cat. No.:
    76484-212
Specifications
About this item
TCEP is a common reducing reagent widely used to break disulfide bonds within and between proteins.
  • Ready to use
  • Simple and effective protocol
Caution: For research use only.