(Ac-KQL)2R110
The non-fluorescent R110 substrates generate the bright green fluorescent rhodamine 110 product that has Ex/Em = 494/521 nm, and can be easily detected with a FITC filter set.
- protease substrate
- bright signal
- multiple applications
- :5,II
- Pack type:Plastic bottle
- Size:1 mg