Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Z-DEVD-FMK

Z-DEVD-FMK

Plastic bottle, 1 mg
Catalog # 76485-298
Irritant
Supplier:   AAT Bioquest
Supplier #:  13303
Z-DEVD-FMK
Catalog # 76485-298
Supplier:  AAT Bioquest
CAS Number:  210344-95-9
Z-DEVD-FMK
Z-DEVD-FMK

Plastic bottle, 1 mg

CAS Number:  210344-95-9
Price
$121.47
Log in for availability
Each (1mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Z-DEVD-FMK is a specific, irreversible Caspase-3 inhibitor, and also shows potent inhibition on caspase-6, caspase-7, caspase-8, and caspase-10.

  • multiple caspases inhibitor
  • enhanced stability
  • increased cell permeability
  • CAS Number:210344-95-9
  • Formula:C₃₀H₄₁FN₄O₁₂
  • MDL Number:MFCD23380217
  • Molecular Weight:668.66 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.