ContactUsLinkComponent
ApoSight™ Green Caspase 3/7 substrate, AAT Bioquest
ApoSight™ Green Caspase 3/7 substrate, AAT Bioquest
Catalog # 76485-690
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ApoSight™ Green Caspase 3/7 substrate, AAT Bioquest
Catalog # 76485-690
Supplier:  AAT Bioquest
Supplier Number:  13290
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pack type:
    Plastic bottle
  • Description:
    ApoSight™ Green Caspase 3/7 substrate
  • Cat. No.:
    76485-690
  • No. of tests:
    100
Specifications
About this item
ApoSight™ Green Caspase 3/7 substrate is the first fluorogenic probe for the direct fluorescence imaing of caspase activities in live cells.
  • unique fluorogenic caspase activity probe for live cells
  • sensitive and accurate detection
  • can be adapted for automation
Caution: Storage: Freeze (< -15 °C)