Order Entry
Puerto Rico
ContactUsLinkComponent
ReadiLink™ Rapid iFluor® 647 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
ReadiLink™ Rapid iFluor® 647 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Catalog # 76481-260
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ReadiLink™ Rapid iFluor® 647 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Catalog # 76481-260
Supplier:  AAT Bioquest
Supplier Number:  1235
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    ReadiLink™ rapid iFluor™ 647 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
  • Size:
    2 labelings
  • Cat. No.:
    76481-260

Specifications

About this item

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Caution: For research use only.