ReadiLink™ Rapid mFluor™ Red 700 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
76481-234
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:ReadiLink™ rapid mFluor™ red 700 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
- Size:2 labelings
- Cat. No.:76481-234
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
- Red-laser excitable
- Robust and simple antibody labeling protocol
- No column purification required
: For research use only.