- Description:ReadiLink™ xtra rapid Cy3 antibody labeling kit 'BSA-compatible'
- Size:2 labelings
- Cat. No.:76485-434
ReadiLink™ xtra rapid antibody labeling kits require essentially only 2 simple mixing steps without a column purification needed.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labeling protocol
- No column purification required