Specifications
- Description:ReadiLink™ xtra rapid iFluor™ 350 antibody labeling kit 'BSA-compatible'
- Size:2 labelings
- Cat. No.:76485-420
Specifications
About this item
ReadiLink™ xtra rapid antibody labeling kits require essentially only two simple mixing steps without a column purification needed.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labeling protocol
- No column purification required