Order Entry
Puerto Rico
ContactUsLinkComponent
ReadiLink™ Rapid Cy7 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
ReadiLink™ Rapid Cy7 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Catalog # 76484-492
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ReadiLink™ Rapid Cy7 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Catalog # 76484-492
Supplier:  AAT BIOQUEST INC
Supplier Number:  1294
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    ReadiLink™ rapid Cy7 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
  • Size:
    2 labelings
  • Cat. No.:
    76484-492

Specifications

About this item

Cy7 is one of the most popular fluorescent labeling dyes for preparing orange-red fluorescent bioconjugates.

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Caution: For research use only.