Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Column Format:Capillary
- Ø int.:12.00 mm
- Capacity:2 ml
- Description:ReadiUse™ bio-gel P-6 spin column
- Length:75 mm
- Cat. No.:76485-064
- Sample volume:30 - 100 µl
Specifications
About this item
ReadiUse™ Spin chromatography columns are packed with polyacrylamide P-6-DG matrices for buffer exchange and desalting applications.
- Exclusion limit of MW 6000 retained
- Exclusion limit above MW 15000 eluted
- swinging bucket centrifuge compatible
- CAS: 25034-58-6