Order Entry
Puerto Rico
ContactUsLinkComponent
Biotin hydrazide, ReadiView™ biodetection
Irritant
Biotin hydrazide, ReadiView™ biodetection
Catalog # 76481-642
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Irritant
Biotin hydrazide, ReadiView™ biodetection
Catalog # 76481-642
Supplier:  AAT Bioquest
Supplier Number:  3055
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    5 mg
  • Cat. No.:
    76481-642

Specifications

About this item

 :
 :258.34 g/mol
ADR:5,II
 :GHS07
CAS Number:66640-86-6
Formula:C₁₀H₁₈N₄O₂S
MDL Number:MFCD00078532