Orders LinkContactUsLinkComponent
Eu succinimidyl ester, trFluor™ fluorescent probe
Eu succinimidyl ester, trFluor™ fluorescent probe
Plastic bottle, 1 mg
  76481-298
Supplier #:  1433
Eu succinimidyl ester, trFluor™ fluorescent probe
  76481-298
 :  
Eu succinimidyl ester, trFluor™ fluorescent probe
Eu succinimidyl ester, trFluor™ fluorescent probe
Plastic bottle, 1 mg
 :  
$646.22
Each (1mg)
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • no crosslinking
  • High stability and selectivity
  • Insensitive to fluorescence quenching
  • High emission yield
  •  :1705.66 g/mol
  •  :5,II
  • Pack type:
    Plastic bottle
  • Size:
    1 mg