Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), IRDye® 800
Catalog # 76485-080
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Vial
- Conjugation:IRDye® 800
- Protein/Peptide Type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Size:200 µg
- Protein Synonyms:Avidin, Streptomyces avidinii
- Protein/Peptide Name:Streptavidin
- Cat. No.:76485-080
Specifications
About this item
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Extremely high biotin-binding affinity
- High quality conjugate
Caution: For research use only.