Order Entry
Puerto Rico
ContactUsLinkComponent
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), FITC (Fluorescein Isothiocyanate)
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), FITC (Fluorescein Isothiocyanate)
Catalog # 76482-712
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), FITC (Fluorescein Isothiocyanate)
Catalog # 76482-712
Supplier:  AAT BIOQUEST INC
Supplier Number:  16910
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/Peptide Type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Size:
    1 mg
  • Protein Synonyms:
    Avidin, Streptomyces avidinii
  • Protein/Peptide Name:
    Streptavidin
  • Cat. No.:
    76482-712

Specifications

About this item

Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.

  • Extremely high biotin-binding affinity
  • High quality conjugate
Caution: For research use only.