ContactUsLinkComponent
Annexin V-iFluor® 700 Conjugate, AAT Bioquest
Catalog # 76483-014
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Annexin V-iFluor® 700 Conjugate, AAT Bioquest
Catalog # 76483-014
Supplier:  AAT Bioquest
Supplier Number:  20077
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Annexin V-iFluor™ 700 conjugate
  • Cat. No.:
    76483-014
  • No. of tests:
    100
Specifications
About this item
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
Caution: For research use only.