Specifications
- Description:Annexin V, XFD594 conjugate
- Cat. No.:76484-566
- No. of tests:100
Specifications
About this item
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- High stability
- In-house quality control
- Multiple research applications