ContactUsLinkComponent
Amplite® Fluorimetric Hypochlorite (Hypochlorous Acid) Assay Kit , AAT Bioquest
Amplite® Fluorimetric Hypochlorite (Hypochlorous Acid) Assay Kit , AAT Bioquest
Catalog # 76482-288
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Amplite® Fluorimetric Hypochlorite (Hypochlorous Acid) Assay Kit , AAT Bioquest
Catalog # 76482-288
Supplier:  AAT Bioquest
Supplier Number:  13846
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    Amplite™ Fluorimetric hypochlorite (hypochlorous acid) assay kit
  • Cat. No.:
    76482-288
  • No. of tests:
    200
Specifications
About this item
Hypochlorite anion (ClO-) and its protonated form, hypochlorous acid (HClO) are critical reactive oxygen species (ROS) in biological systems.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.